Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 5-HT7 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10325T100UG
This item is not returnable.
View return policy
Description
5-HT7 Monoclonal specifically detects 5-HT7 in Human samples. It is validated for Flow Cytometry.Specifications
5-HT7 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
5HT7, 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), HTR7, serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
Synthetic peptide containing human 5-HTR7, Accession # P34969 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1015322 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P34969 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3363 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction