Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5-HT7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10325100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10325100 100 μg
MAB10325SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10325100 Supplier R&D Systems Supplier No. MAB10325100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

5-HT7 Monoclonal specifically detects 5-HT7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen 5-HT7
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 1015322
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. P34969
Gene Alias 5HT7, 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), HTR7, serotonin 5-HT-7 receptor, Serotonin receptor 7
Host Species Mouse
Immunogen Synthetic peptide containing human 5-HTR7, Accession # P34969
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3363
Test Specificity Detects human 5-HTR7 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.