Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human 5'-Nucleotidase/CD73 ELISA Kit

Catalog No. p-7219718
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Human 5'-Nucleotidase/CD73 quantitates human 5'-Nucleotidase/CD73 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human 5'-Nucleotidase/CD73.

CD73 (Ecto-5-prime-nucleotidase, 5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. CD73 consists of a dimer of 2 identical 70 kDa subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. CD73 is used as a marker of lymphocyte differentiation. Consequently, a deficiency of CD73 occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from CD73 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. The CD73 gene has been localized to 6q14-q21 by immunofluorescence and a study of a panel of human x mouse hybrids that contained fragments of chromosome 6 as translocations. Defects in the CD73 gene can lead to the calcification of joints and arteries, and intestinal tuberculosis. Two transcript variants encoding different isoforms of CD73 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2210401F01Rik; 5 nucleotidase, ecto; 5 ' nucleotidase, ecto; 5 '-NT; 5 '-nucleotidase; 5 '-nucleotidase ecto; 5 '-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; Ecto-5 '-nucleotidase; eN; eNT; NT; Nt5; Nt5e; Nte; Purine 5-Prime-Nucleotidase
Assay Range 35 to 8,000 pg/mL
Assay Sensitivity 35 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4907
Gene Symbol NT5E
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD73
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.