Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 53BP1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$188.35 - $532.11
Specifications
| Antigen | 53BP1 |
|---|---|
| Clone | 1285C |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 3-25 ug/mL, Knockout Validated 2 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18772SP
![]() |
R&D Systems
MAB18772SP |
25 μg |
Each for $188.35
|
|
|||||
MAB18772100
![]() |
R&D Systems
MAB18772100 |
100 μg |
Each for $532.11
|
|
|||||
Description
53BP1 Monoclonal specifically detects 53BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.Specifications
| 53BP1 | |
| Western Blot 2 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 3-25 ug/mL, Knockout Validated 2 ug/mL | |
| Monoclonal | |
| Rabbit | |
| Human | |
| Q12888 | |
| 7158 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1285C | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, TP53BP1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| E. coli-derived recombinant human 53BP1 Ala1614-His1972 Accession # Q12888 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human 53BP1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title