Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 5T4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB49751G100UG
This item is not returnable.
View return policy
Description
5T4 Monoclonal specifically detects 5T4 in Human samples. It is validated for Flow Cytometry.Specifications
5T4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q13641 | |
TPBG | |
Mouse myeloma cell line NS0-derived recombinant human 5T4 Ser31-Ser355 Accession # Q13641 | |
RUO | |
7162 | |
Human | |
IgG1 |
Flow Cytometry | |
524744 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, M6P1, TPBG, trophoblast glycoprotein | |
Mouse | |
100 μg | |
Primary | |
Detects human 5T4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse 5T4 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction