Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human 5T4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB49751
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB49751 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB49751 Supplier R&D Systems Supplier No. MAB49751
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

5T4 Monoclonal specifically detects 5T4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen 5T4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 524744
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13641
Gene Alias 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, M6P1, TPBG, trophoblast glycoprotein
Gene Symbols TPBG
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human 5T4 Ser31-Ser355 Accession # Q13641
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7162
Test Specificity Detects human 5T4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse 5T4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.