Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human A20/TNFAIP3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7598SP
This item is not returnable.
View return policy
Description
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human samples. It is validated for Western Blot.Specifications
A20/TNFAIP3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P21580 | |
TNFAIP3 | |
E. coli-derived recombinant human A20 Lys91-Leu263 Accession # P21580 | |
25 μg | |
Primary | |
Detects human A20 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human A20/TNFAIP3 (aa 440-790) is observed. | |
Human | |
Purified |
Western Blot | |
775912 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, TNFA1P2, TNFAIP3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7128 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction