Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human A33 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB30804U100UG
This item is not returnable.
View return policy
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Flow CytometrySpecifications
A33/GPA33 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), GPA33, MGC129986, MGC129987, transmembrane glycoprotein A33 | |
E. coli-derived recombinant human A33, Ile22-Val235, Accession # Q99795 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2963F | |
Flow Cytometry | |
Q99795 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10223 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction