Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AAK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6886SP
This item is not returnable.
View return policy
Description
AAK1 Monoclonal specifically detects AAK1 in Human samples. It is validated for Western Blot.Specifications
AAK1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q2M2I8 | |
AAK1 | |
E. coli-derived recombinant human AAK1 Ser704-Ile822 Accession # Q2M2I8 | |
25 μg | |
Primary | |
Detects human AAK1 in direct ELISAs. | |
Human | |
Purified |
Western Blot | |
702425 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22848 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction