Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AAK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6886
This item is not returnable.
View return policy
Description
AAK1 Monoclonal specifically detects AAK1 in Human samples. It is validated for Western Blot.Specifications
AAK1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q2M2I8 | |
AAK1 | |
E. coli-derived recombinant human AAK1 Ser704-Ile822 Accession # Q2M2I8 | |
100 μg | |
Primary | |
Detects human AAK1 in direct ELISAs. | |
Human | |
Purified |
Western Blot | |
702425 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22848 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction