Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AATK Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4866V100UG
This item is not returnable.
View return policy
Description
AATK/Serine/threonine-protein kinase LMTK1 Monoclonal specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| AATK/Serine/threonine-protein kinase LMTK1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAH47378 | |
| AATK | |
| E. coli-derived recombinant human AATK Asn421-Ala589 Accession # AAH47378 | |
| RUO | |
| 9625 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 456006 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AATYK, AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1, LMR1EC 2.7.11.1, LMTK1, LMTK1AATYK1, p35BP, serine/threonine-protein kinase LMTK1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human AATK in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction