Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ABCA1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB72071100
This item is not returnable.
View return policy
Description
ABCA1 Monoclonal specifically detects ABCA1 in Human samples. It is validated for Western Blot.Specifications
| ABCA1 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ABC1, ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGD, TGDATP-binding cassette sub-family A member 1 | |
| Human ABCA1 synthetic peptide Accession # O95477 | |
| 100 μg | |
| Primary | |
| Detects human ABCA1 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 1276B | |
| Western Blot 1 ug/mL | |
| O95477 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 19 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction