Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ABCA13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11195SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11195SP 25 μg
MAB11195100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11195SP Supplier R&D Systems Supplier No. MAB11195SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ABCA13 Monoclonal antibody specifically detects ABCA13 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen ABCA13
Applications Immunohistochemistry
Classification Monoclonal
Clone 2379B
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q86UQ4
Gene Alias ATP binding cassette transporter A13, ATP-binding cassette sub-family A member 13, ATP-binding cassette, sub-family A (ABC1), member 13, DKFZp313D2411, EC 2.7.11.25, EC 3.6.3, FLJ16398, FLJ33876, FLJ33951
Host Species Rabbit
Immunogen Synthetic peptide within aminoacids 4700-4750 of the Human ABCA13 protein,, Accession # Q86UQ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154664
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.