Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ABCE1 Full-length ORF (NP_001035809.1, 1 a.a. - 599 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7096213
Click to view available options
Quantity:
10 ug

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. Alternatively referred to as the RNase L inhibitor, this protein functions to block the activity of ribonuclease L. Activation of ribonuclease L leads to inhibition of protein synthesis in the 2-5A/RNase L system, the central pathway for viral interferon action. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MADKLTRIAIVNHDKCKPKKCRQECKKSCPVVRMGKLCIEVTPQSKIAWISETLCIGCGICIKKCPFGALSIVNLPSNLEKETTHRYCANAFKLHRLPIPRPGEVLGLVGTNGIGKSTALKILAGKQKPNLGKYDDPPDWQEILTYFRGSELQNYFTKILEDDLKAIIKPQYVDQIPKAAKGTVGSILDRKDETKTQAIVCQQLDLTHLKERNVEDLSGGELQRFACAVVCIQKADIFMFDEPSSYLDVKQRLKAAITIRSLINPDRYIIVVEHDLSVLDYLSDFICCLYGVPSAYGVVTMPFSVREGINIFLDGYVPTENLRFRDASLVFKVAETANEEEVKKMCMYKYPGMKKKMGEFELAIVAGEFTDSEIMVMLGENGTGKTTFIRMLAGRLKPDEGGEVPVLNVSYKPQKISPKSTGSVRQLLHEKIRDAYTHPQFVTDVMKPLQIENIIDQEVQTLSGGELQRVALALCLGKPADVYLIDEPSAYLDSEQRLMAARVVKRFILHAKKTAFVVEHDFIMATYLADRVIVFDGVPSKNTVANSPQTLLAGMNKFLSQLEITFRRDPNNYRPRINKLNSIKDVEQKKSGNYFFLDD

Specifications

Accession Number NP_001035809.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6059
Molecular Weight (g/mol) 93.7kDa
Name ABCE1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ABC38/OABP/RLI/RNASEL1/RNASELI/RNS4I
Common Name ABCE1
Gene Symbol ABCE1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.