Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ABCG2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB995P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB995P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB995P Supplier R&D Systems Supplier No. FAB995P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ABCG2/CD338
Applications Flow Cytometry
Classification Monoclonal
Clone 5D3
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias ABC transporter, ABC15, ABCP, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXR, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Gene Symbols ABCG2
Host Species Mouse
Immunogen 3T3 cells transduced with human ABCG2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9429
Test Specificity Detects human ABCG2 in flow cytometry and immunocytochemistry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.