Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ACE-2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB933T100UG
This item is not returnable.
View return policy
Description
ACE-2 Polyclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.Specifications
ACE-2 | |
Polyclonal | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BYF1 | |
Goat | |
Antigen Affinity-purified | |
RUO | |
59272 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction