Learn More
Human ACE-2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems FAB933R100UG
Catalog No. FAB933R100U
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
ACE-2 Polyclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ACE-2
Polyclonal
Flow Cytometry 0.25-1 ug/10^6 cells
Q9BYF1
Goat
Antigen Affinity-purified
RUO
59272
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1
100 μg
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.