Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ACE-2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | ACE-2 |
---|---|
Clone | 2817M |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 0.3-25 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10823SP
![]() |
R&D Systems
MAB10823SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10823100
![]() |
R&D Systems
MAB10823100 |
100 μg |
Each for $479.50
|
|
|||||
Description
ACE-2 Monoclonal antibody specifically detects ACE-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
ACE-2 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 0.3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
Mouse myeloma cell line NS0-derived human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2817M | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9BYF1 | |
59272 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title