Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ACE/CD143 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM929
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM929 Supplier R&D Systems Supplier No. BAM929
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ACE/CD143
Applications Flow Cytometry
Classification Monoclonal
Clone 171417
Concentration LYOPH
Conjugate Biotin
Dilution Flow Cytometry 2.5 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P12821
Gene Alias ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Gene Symbols ACE
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ACE/CD143 Leu30-Leu1261 Accession # P12821
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1636
Test Specificity Detects human ACE in direct ELISAs. Does not cross-react with recombinant human ACE-2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.