Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ACE/CD143 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB929P
This item is not returnable.
View return policy
Description
ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Flow Cytometry.Specifications
ACE/CD143 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ACE | |
Mouse myeloma cell line NS0-derived recombinant human ACE/CD143 aa 30-1261 | |
100 Tests | |
Primary | |
Detects human ACE/CD143 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ACE-2 is observed. Detects the surface expression of human ACE on full length ACE transfectants, but not on control transfectants by flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
171417 | |
Flow Cytometry 10 uL/10^6 cells | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1636 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction