Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Acetyl Histone H4 (Lys5) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$164.60 - $343.65
Specifications
| Antigen | Histone H4 |
|---|---|
| Clone | 2129F |
| Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 0.1-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9549SP
![]() |
R&D Systems
MAB9549SP |
25 μg |
Each for $164.60
|
|
|||||
MAB9549100
![]() |
R&D Systems
MAB9549100 |
100 μg |
Each for $343.65
|
|
|||||
Description
Histone H4 Monoclonal specifically detects Histone H4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Histone H4 | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 0.1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| H4, H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4M, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, HIST4H4, histone 4, H4, Histone Cluster 1 H4, Histone Cluster 1 H4i, Histone Cluster 4 H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Human Histone H4 synthetic peptide Accession # P62805 (H4_Human) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Acetyl Histone H4 when acetylated at Lys5 in Western blots. |
| 2129F | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P62805 (H4_Human) | |
| 121504 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title