Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Acetylated p53 (K382) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB13552100
This item is not returnable.
View return policy
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
p53 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Synthetic peptide containing acetylated human p53 (K382), Accession # P04637 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
2589A | |
Western Blot 4 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
P04637 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
7157 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction