Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Acetylcholinesterase/ACHE Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. p-7121598 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7574SP 25 μg
AF7574100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF7574SP Supplier R&D Systems Supplier No. AF7574SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Acetylcholinesterase/ACHE
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 3-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P22303
Gene Alias acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43
Test Specificity Detects human ACHE in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.