Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ACLP Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6425N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6425N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6425N100 Supplier R&D Systems Supplier No. IC6425N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AE Binding Protein 1/ACLP Monoclonal specifically detects AE Binding Protein 1/ACLP in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen AE Binding Protein 1/ACLP
Applications Flow Cytometry
Classification Monoclonal
Clone 590831
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8IUX7
Gene Alias ACLP, ACLPadipocyte enhancer binding protein 1, AE binding protein 1, AE-binding protein 1adipocyte enhancer-binding protein 1, AEBP1, Aortic carboxypeptidase-like protein, FLJ33612
Gene Symbols AEBP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ACLP Glu345-Phe1158 Accession # Q8IUX7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165
Test Specificity Detects human ACLP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ACLP is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.