Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ACLP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6425SP
This item is not returnable.
View return policy
Description
AE Binding Protein 1/ACLP Monoclonal specifically detects AE Binding Protein 1/ACLP in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
AE Binding Protein 1/ACLP | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q8IUX7 | |
AEBP1 | |
Mouse myeloma cell line NS0-derived recombinant human ACLP Glu345-Phe1158 Accession # Q8IUX7 | |
25 μg | |
Primary | |
Detects human ACLP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ACLP is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
590831 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACLP, ACLPadipocyte enhancer binding protein 1, AE binding protein 1, AE-binding protein 1adipocyte enhancer-binding protein 1, AEBP1, Aortic carboxypeptidase-like protein, FLJ33612 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
165 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction