Learn More
Description
Specifications
Specifications
| Assay Range | 1.56 - 100 ng/mL |
| Assay Sensitivity | 0.94 ng/mL |
| Cross Reactivity | This kit recognizes Human AAA in samples. No Significant cross-reactivity or interference between Human AAA and analogues was observed. |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 58 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
