Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Active MMP-9 Fluorokine E Kit
SDP

Catalog No. F9M00 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F9M00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F9M00 Supplier R&D Systems™ Supplier No. F9M00
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains Mercury, Dispose According to Local, State or Federal Laws.

The Fluorokine™ E Human Active MMP-9 Florescent Assay combines the specificity of a monoclonal antibody that captures all three MMP-9 forms (92, 82, and 65 kDa) but not other MMPs and the sensitivity of fluorescence. The assay is designed to measure the levels of endogenous human active MMP-9 in cell culture supernates, serum, plasma, and urine and the MMP-9 in these samples that can be activated by APMA during the assay procedure. The determined MMP-9 activity may reflect the balance of MMP-9 and its inhibitor TIMPs. Therefore, it is recommended that the levels of TIMPs be determined in order to interpret the results properly. For example, TIMP-1 inhibits the activity of MMP-9 through binding of the N-terminal domain of the TIMP-1 to the active site of MMP-9. In addition, TIMP-1 also binds to pro-MMP-9 through the interaction between the C-terminal domains of both proteins .

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detection Method Fluorogenic
Gene ID (Entrez) 4318
Gene Symbol MMP9
Product Line R&D Systems
Quantity 1 Kit
Synonym 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Type Activity Assay Kit
Target MMP-9
Assay Range 0.25 to 16 pg/mL
Assay Sensitivity 0.01 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.