Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Activin B beta B subunit Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB659500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB659500 500 μg
MAB659SP 25 μg
MAB659100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB659500 Supplier R&D Systems Supplier No. MAB659500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Activin B/Inhibin beta B Monoclonal specifically detects Activin B/Inhibin beta B in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Activin B/Inhibin beta B
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 146807
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.6-3.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q53T31
Gene Alias activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2
Gene Symbols INHBB
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Activin B Gly293-Ala407 Accession # Q53T31
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3625
Test Specificity Detects human Activin B beta B subunit in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Inhibin B or rhActivin A is observed. In Western blots, approximately 20% cross-reactivity with rhInhibin B is observed and no cross-reactivity with rhActivin A or recombinant mouse Activin A is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.