Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin C/Inhibin beta C Antibody, Unconjugated, R&D Systems™
GREENER_CHOICE

Catalog No. AF1629
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1629 100 μg
AF1629SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1629 Supplier R&D Systems Supplier No. AF1629
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Activin C/Inhibin beta C Polyclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Activin C/Inhibin beta C
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA
Gene Accession No. 3626
Gene Alias inhibin beta C chain, IHBC, INHBC, inhibin, beta C
Gene Symbols INHBC
Host Species Goat
Immunogen E. coli-derived recombinant human Activin C Gly237-Ser352 Accession # 3626
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3626
Test Specificity Detects human Activin C beta C subunit in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.