Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIA/ALK-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF637
This item is not returnable.
View return policy
Description
Activin RIA/ALK-2/Activin Receptor Type 1 Polyclonal specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Activin RIA/ALK-2/Activin Receptor Type 1 | |
Polyclonal | |
Unconjugated | |
Q04771 | |
ACVR1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Activin RIA/ALK-2 Asp23-Glu123 Accession # Q04771 | |
100 μg | |
Primary | |
Detects human Activin RIA/ALK-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Activin RIB, rhActivin RIIA, and rhActivin RIIB is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
activin A receptor, type I, ActivinRIA, ACTRI, ACVR1, ACVR1A, ACVRLK2, ALK2, ALK-2, FOP, SKR1, TSRI | |
Goat | |
Affinity Purified | |
RUO | |
90 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction