Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIB/ALK-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB222
This item is not returnable.
View return policy
Description
Activin RIB/ALK-4 Monoclonal specifically detects Activin RIB/ALK-4 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
Activin RIB/ALK-4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
91 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Blockade of Receptor-ligand Interaction | |
126822 | |
Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction | |
AAA60556 | |
ACVR1B | |
Mouse myeloma cell line NS0-derived recombinant human Activin RIB/ALK-4 Ser24-Glu126 Accession # AAA60556 | |
500 μg | |
Primary | |
Detects human Activin RIB/ALK-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Activin RIA, rhActivin RIIA, or rhActivin RIIB. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction