Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Activin RIB/ALK-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB222SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB222SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB222SP Supplier R&D Systems Supplier No. MAB222SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Activin RIB/ALK-4 Monoclonal specifically detects Activin RIB/ALK-4 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen Activin RIB/ALK-4
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 126822
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA60556
Gene Alias activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B
Gene Symbols ACVR1B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Activin RIB/ALK-4 Ser24-Glu126 Accession # AAA60556
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91
Test Specificity Detects human Activin RIB/ALK-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Activin RIA, rhActivin RIIA, or rhActivin RIIB.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.