Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Activin RIIA/B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3391
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3391 500 μg
MAB3391SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3391 Supplier R&D Systems Supplier No. MAB3391
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Activin RIIA/B Monoclonal specifically detects Activin RIIA/B in Human samples. It is validated for Western Blot.

Specifications

Antigen Activin RIIA/B
Applications Western Blot
Classification Monoclonal
Clone 60401
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13705
Gene Alias ActivinRIIA, ActivinRIIB, ACTRII, ACVR2, ACVR2A, ACVR2B, AVR2A
Gene Symbols ACVR2A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Activin RIIB Ser19-Thr134 Accession # Q13705
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92
Test Specificity Detects human Activin RII in direct ELISA and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.