Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Activin RIIA Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems BAF340
This item is not returnable.
View return policy
Description
Activin RIIA Polyclonal specifically detects Activin RIIA in Human samples. It is validated for Western Blot.Specifications
Activin RIIA | |
Polyclonal | |
Biotin | |
P27037 | |
ACVR2A | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Activin RIIA Ser25-Asp82 Accession # P27037 | |
50 μg | |
Primary | |
Detects human ActivináRIIA in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ActivinRIIA, ACTRII, ACTRIIA, ACVR2A, ACVR2ACTR-IIA, AVR2A, EC 2.7.11, EC 2.7.11.30 | |
Goat | |
Affinity Purified | |
RUO | |
92 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction