Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human Activin RIIB Antibody, R&D Systems™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$200.50 - $1973.00
Specifications
| Antigen | Activin RIIB | 
|---|---|
| Clone | 60408 | 
| Dilution | Western Blot 1 ug/mL | 
| Applications | Western Blot | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| MAB3392SP  | R&D Systems MAB3392SP | 25 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $200.50
                                                 |  | |||||
| MAB3392100  | R&D Systems MAB3392100 | 100 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $538.00
                                                 |  | |||||
| MAB3392500  | R&D Systems MAB3392500 | 500 μg | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $1,973.00
                                                 |  | |||||
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot.Specifications
| Activin RIIB | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| CAA54671 | |
| 93 | |
| Mouse myeloma cell line NS0-derived recombinant human Activin RIIB Ser19-Thr134 Accession # CAA54671 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Activin RIIB in direct ELISAs and Western blots. Shows approximately 5% cross-reactivity with recombinant human Activin RIIA. | 
| 60408 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActivinRIIB, ActR-IIB, ACTRIIB, ACVR2B, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| ACVR2B | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            