Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAM10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $538.00
Specifications
| Antigen | ADAM10 |
|---|---|
| Clone | 172023 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14271SP
![]() |
R&D Systems
MAB14271SP |
25 μg |
Each for $214.00
|
|
|||||
MAB14271100
![]() |
R&D Systems
MAB14271100 |
100 μg |
Each for $538.00
|
|
|||||
Description
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for ELISA.Specifications
| ADAM10 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| O14672 | |
| 102.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 172023 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human ADAM10 protein, Thr214-Glu672, with a C-terminal 10-His tag, Accession No. O14672 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ADAM10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title