Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB44161SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB44161SP 25 μg
MAB44161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB44161SP Supplier R&D Systems Supplier No. MAB44161SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen ADAM12
Applications Immunoprecipitation
Classification Monoclonal
Clone 632525
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43184
Gene Alias a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.-
Gene Symbols ADAM12
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # O43184
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8038
Test Specificity Detects human ADAM12 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ADAM8, 9, 10, 15, 17, 19, 22, 23, 32, or 33 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.