Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAM12 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4416
This item is not returnable.
View return policy
Description
ADAM12 Polyclonal specifically detects ADAM12 in Human samples. It is validated for Western Blot.Specifications
ADAM12 | |
Polyclonal | |
Biotin | |
AAC08702 | |
ADAM12 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # AAC08702 | |
50 μg | |
Primary | |
Detects human ADAM12 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) ADAM8 is observed and less than 1% cross-reactivity with rhADAM19 and rhADAM33 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
Sheep | |
Affinity Purified | |
RUO | |
8038 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction