Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM15 Ectodomain Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC935G100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC935G100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC935G100UG Supplier R&D Systems Supplier No. IC935G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAM15 Monoclonal specifically detects ADAM15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ADAM15
Applications Flow Cytometry
Classification Monoclonal
Clone 23G9
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q13444
Gene Alias ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin
Gene Symbols ADAM15
Host Species Mouse
Immunogen COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant human ADAM15 Asp207-Thr696 Accession # Q13444
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8751
Test Specificity Detects the ectodomain of recombinant human (rh) ADAM15 in direct ELISAs and Western blots. In direct ELISAs, shows 100% cross-reactivity with recombinant mouse (rm) ADAM15 but does not cross-react with rhADAM8, 9, 17, 28, rmADAM9, or 10.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.