Learn More
Human ADAM15 Ectodomain Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ADAM15 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
| Gene Accession No. | Q13444 |
| Gene Alias | ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin |
| Gene Symbols | ADAM15 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.