Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM22 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB49081SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB49081SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB49081SP Supplier R&D Systems Supplier No. MAB49081SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAM22 Monoclonal specifically detects ADAM22 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ADAM22
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 498805
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9P0K1
Gene Alias ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832
Gene Symbols ADAM22
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 Accession # Q9P0K1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53616
Test Specificity Detects human ADAM22 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ADAM8, 9, 10, 12, 15, 19, 23, 32, or 33 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.