Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM28 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB93252100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB93252100 100 μg
MAB93252SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB93252100 Supplier R&D Systems Supplier No. MAB93252100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAM28 Monoclonal specifically detects ADAM28 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen ADAM28
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1004266
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q0UKQ2
Gene Alias a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, Dtgn1, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCL, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L, Protein-L
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 & Leu191-Ala623,, Accession # Q0UKQ2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10863
Test Specificity Detects human ADAM28 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.