Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAM28 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$176.44 - $449.61
Specifications
| Antigen | ADAM28 |
|---|---|
| Clone | 2248A |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9325SP
![]() |
R&D Systems
MAB9325SP |
25 μg |
Each for $176.44
|
|
|||||
MAB9325100
![]() |
R&D Systems
MAB9325100 |
100 μg |
Each for $449.61
|
|
|||||
Description
ADAM28 Monoclonal specifically detects ADAM28 in Human samples. It is validated for ELISA.Specifications
| ADAM28 | |
| ELISA | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, Dtgn1, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCL, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L, Protein-L | |
| Chinese Hamster Ovary cell line, CHO-derived human ADAM28 protein, Leu72 & Leu191-Ala623, with a C-terminal 6-His tag, Accession No. Q0UKQ2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ADAM28 in direct ELISAs. |
| 2248A | |
| ELISA | |
| Unconjugated | |
| Human | |
| Q0UKQ2 | |
| 10863.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title