Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM32 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5380
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5380 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5380 Supplier R&D Systems Supplier No. BAF5380
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ADAM32 Polyclonal specifically detects ADAM32 in Human samples. It is validated for Western Blot.

Specifications

Antigen ADAM32
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q8TC27
Gene Alias a disintegrin and metalloprotease domain 32, a disintegrin and metalloproteinase domain 32, ADAM 32, ADAM metallopeptidase domain 32, disintegrin and metalloproteinase domain-containing protein 32, FLJ26299, FLJ29004, metalloproteinase 12-like protein
Gene Symbols ADAM32
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAM32 Ser17-Thr476 Accession # Q8TC27
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203102
Test Specificity Detects human ADAM32 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) ADAM9, rhADAM22, and rhADAM23 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.