Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAM33 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5565S100UG
This item is not returnable.
View return policy
Description
ADAM33 Monoclonal specifically detects ADAM33 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
ADAM33 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BZ11 | |
ADAM33 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM33 Val30-Leu501 (predicted) Accession # Q9BZ11 | |
RUO | |
80332 | |
Human | |
IgG1 |
Flow Cytometry | |
508101 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
Mouse | |
100 μg | |
Primary | |
Detects human ADAM33 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse ADAM33 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction