Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAMTS1 Partial ORF (AAH36515, 858 a.a. - 967 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098096
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two disintegrin loops and three C-terminal TS motifs and has anti-angiogenic activity. The expression of this gene may be associated with various inflammatory processes as well as development of cancer cachexia. This gene is likely to be necessary for normal growth, fertility, and organ morphology and function. [provided by RefSeq]

Sequence: WVIEEWGECSKSCELGWQRRLVECRDINGQPASECAKEVKPASTRPCADHPCPQWQLGEWSSCSKTCGKGYKKRSLKCLSHDGGVLSHESCDPLKKPKHFIDFCTMAECS

Specifications

Accession Number AAH36515
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9510
Molecular Weight (g/mol) 37.84kDa
Name ADAMTS1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen WVIEEWGECSKSCELGWQRRLVECRDINGQPASECAKEVKPASTRPCADHPCPQWQLGEWSSCSKTCGKGYKKRSLKCLSHDGGVLSHESCDPLKKPKHFIDFCTMAECS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C3-C5/KIAA1346/METH1
Common Name ADAMTS1
Gene Symbol ADAMTS1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.