Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS1 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3079
This item is not returnable.
View return policy
Description
ADAMTS1 Polyclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
ADAMTS1 | |
Polyclonal | |
Unconjugated | |
Q9UHI8 | |
ADAMTS1 | |
Mouse myeloma cell line NS0-derived recombinant human ADAMTS1 Leu50-Arg252 Accession # Q9UHI8 | |
100 μg | |
Primary | |
Detects human ADAMTS1 Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with mature recombinant human ADAMTS1 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH1, METH-1 | |
Goat | |
Affinity Purified | |
RUO | |
9510 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction