Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5270
This item is not returnable.
View return policy
Description
ADAMTS10 Monoclonal specifically detects ADAMTS10 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAMTS10 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q9H324 | |
ADAMTS10 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS10 Phe26-Gly1102 Accession # Q9H324 | |
100 μg | |
Primary | |
Detects human ADAMTS10 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human ADAMTS1, 3, 4, 5, 12, 13, 15, 16, or L2 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
496708 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 10, ADAM metallopeptidase with thrombospondin type 1 motif, 10, ADAM-TS 10, ADAMTS-10, ADAM-TS10A disintegrin and metalloproteinase with thrombospondin motifs 10, EC 3.4.24.-, EC 3.4.24.82, WMS, zinc metalloendopeptidase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81794 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction