Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $464.50
Specifications
Antigen | ADAMTS13 |
---|---|
Clone | 442315 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42451100
![]() |
R&D Systems
MAB42451100 |
100 μg |
Each for $464.50
|
|
|||||
MAB42451SP
![]() |
R&D Systems
MAB42451SP |
25 μg |
Each for $177.10
|
|
|||||
Description
ADAMTS13 Monoclonal antibody specifically detects ADAMTS13 in Human samples. It is validated for ELISASpecifications
ADAMTS13 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTP, TTPADAM-TS 13, vWF-cleaving protease, vWF-CP, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS13,, Gln34-Trp688, Accession # Q76LX8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
442315 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q76LX8 | |
11093 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title