Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ADAMTS13 Partial ORF (NP_620594, 1328 a.a. - 1427 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099097
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: FINVAPHARIAIHALATNMGAGTEGANASYILIRDTHSLRTTAFHGQQVLYWESESSQAEMEFSEGFLKAQASLRGQYWTLQSWVPEMQDPQSWKGKEGT

Specifications

Accession Number NP_620594
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11093
Molecular Weight (g/mol) 36.74kDa
Name ADAMTS13 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen FINVAPHARIAIHALATNMGAGTEGANASYILIRDTHSLRTTAFHGQQVLYWESESSQAEMEFSEGFLKAQASLRGQYWTLQSWVPEMQDPQSWKGKEGT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C9orf8/DKFZp434C2322/FLJ42993/MGC118899/MGC118900/TTP/VWFCP/vWF-CP
Common Name ADAMTS13
Gene Symbol ADAMTS13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.